Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdbf2
Entrez Gene ID
501153
Gene Name
zinc finger, DBF-type containing 2
Gene Aliases
RGD1560005
Chromosome Location
Chr.9: 70052087 - 70090847 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245044.2 XP_006245106.1 2 - 3 142 77
XM_006245045.2 XP_006245107.1 2 - 3 125 77
XM_006245046.2 XP_006245108.1 2 - 3 172 77

View Full Product Details