Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sumo1
Entrez Gene ID
301442
Gene Name
small ubiquitin-like modifier 1
Gene Aliases
-
Chromosome Location
Chr.9: 66453428 - 66483614 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ214389.1 - 1 - 2 138 130
FQ214927.1 - 1 - 2 121 130
FQ215001.1 - 1 - 2 131 130
FQ224870.1 - 1 - 2 120 130
FQ234204.1 - 1 - 2 123 130

View Full Product Details