Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyg1
Entrez Gene ID
503254
Gene Name
lysozyme G-like 1
Gene Aliases
RGD1560332
Chromosome Location
Chr.9: 44527596 - 44539320 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098838.1 NP_001092308.1 - 100 86

Gene Symbol
LOC100910070
Entrez Gene ID
100910070
Gene Name
lysozyme g-like protein 1-like
Gene Aliases
-
Chromosome Location
Chr.9: 44605450 - 44611774 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006244868.1 XP_006244930.1 1 - 2 42 86

View Full Product Details