The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199113.1 | NP_954544.1 | 2 - 3 | 1030 | 116 |
XM_008768733.1 | XP_008766955.1 | 3 - 4 | 287 | ||
XM_008768734.1 | XP_008766956.1 | 3 - 4 | 287 | ||
GenBank mRNA | AY490241.1 | - | 2 - 3 | 993 | 116 |
BC061876.1 | - | 2 - 3 | 1030 | 116 |