Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss42
Entrez Gene ID
301027
Gene Name
protease, serine, 42
Gene Aliases
RGD1562548, Tessp2
Chromosome Location
Chr.8: 119064961 - 119070160 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106863.2 NP_001100333.2 1 - 2 222 72

View Full Product Details