The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008328.1 | NP_001008329.1 | 9 - 10 | 1376 | 84 |
XM_006243725.2 | XP_006243787.1 | 9 - 10 | 1712 | 84 | |
XM_008766500.1 | XP_008764722.1 | 9 - 10 | 1743 | 84 | |
GenBank mRNA | BC085863.1 | - | 9 - 10 | 1376 | 84 |