The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107290.1 | NP_001100760.1 | 10 - 11 | 1037 | 71 |
XM_006252603.2 | XP_006252665.1 | 10 - 11 | 1019 | 71 | |
GenBank mRNA | BC168886.1 | - | 6 - 7 | 582 | 71 |