Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfe2l1
Entrez Gene ID
360610
Gene Name
nuclear factor, erythroid 2-like 1
Gene Aliases
-
Chromosome Location
Chr.10: 84686762 - 84699301 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC169095.1 - - 2009 84

View Full Product Details