Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wfs1
Entrez Gene ID
83725
Gene Name
Wolfram syndrome 1 (wolframin)
Gene Aliases
-
Chromosome Location
Chr.14: 78640707 - 78665224 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031823.1 NP_114011.1 6 - 7 887 61
GenBank mRNA AF136378.1 - 6 - 7 887 61

View Full Product Details