Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr74
Entrez Gene ID
690229
Gene Name
WD repeat domain 74
Gene Aliases
-
Chromosome Location
Chr.1: 224933913 - 224938659 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109569.1 NP_001103039.1 6 - 7 656 118
XM_006230999.2 XP_006231061.1 6 - 7 663 118
XM_006231000.2 XP_006231062.1 6 - 7 531 118
GenBank mRNA BC158834.1 - 6 - 7 625 118

View Full Product Details