Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wee2
Entrez Gene ID
502750
Gene Name
WEE1 homolog 2 (S. pombe)
Gene Aliases
-
Chromosome Location
Chr.4: 68605097 - 68631299 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762866.1 XP_008761088.1 9 - 10 4311 65

View Full Product Details