Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsf5
Entrez Gene ID
688111
Gene Name
heat shock transcription factor family member 5
Gene Aliases
-
Chromosome Location
Chr.10: 74909581 - 74954124 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768238.1 XP_008766460.1 5 - 6 2277 103

View Full Product Details