Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A1bg
Entrez Gene ID
140656
Gene Name
alpha-1-B glycoprotein
Gene Aliases
C44
Chromosome Location
Chr.7: 102294805 - 102298522 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC089771.1 - 1 - 2 48 94

View Full Product Details