Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb10
Entrez Gene ID
80338
Gene Name
zinc finger and BTB domain containing 10
Gene Aliases
Rinzf
Chromosome Location
Chr.2: 94692939 - 94730976 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024489.1 NP_077815.1 6 - 7 2883 145
GenBank mRNA AF091457.1 - 6 - 7 2883 145

View Full Product Details