Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a11
Entrez Gene ID
287796
Gene Name
solute carrier family 39, member 11
Gene Aliases
-
Chromosome Location
Chr.10: 101754842 - 102169099 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013042.1 NP_001013060.1 4 - 5 418 100
XM_006247663.2 XP_006247725.1 4 - 5 716 100
XM_006247664.2 XP_006247726.1 4 - 5 787 100
XM_006247665.2 XP_006247727.1 4 - 5 716 100
XM_006247668.2 XP_006247730.1 4 - 5 483 100
XM_006247670.2 XP_006247732.1 4 - 5 1267 100
XM_006247671.2 XP_006247733.1 4 - 5 866 100
GenBank mRNA BC062054.1 - 4 - 5 418 100

View Full Product Details