Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrpl33
Entrez Gene ID
100363539
Gene Name
mitochondrial ribosomal protein L33
Gene Aliases
-
Chromosome Location
Chr.6: 26130278 - 26138493 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729600.3 XP_002729646.1 3 - 4 221 150
GenBank mRNA FQ224256.1 - 3 - 4 182 150

View Full Product Details