Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fscn2
Entrez Gene ID
303741
Gene Name
fascin actin-bundling protein 2, retinal
Gene Aliases
-
Chromosome Location
Chr.10: 109528330 - 109540026 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107072.1 NP_001100542.1 3 - 4 1273 94

View Full Product Details