Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dus4l
Entrez Gene ID
366593
Gene Name
dihydrouridine synthase 4-like (S. cerevisiae)
Gene Aliases
RGD1311445
Chromosome Location
Chr.6: 50929156 - 50943488 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135803.1 NP_001129275.1 1 - 2 145 92

View Full Product Details