The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135607.1 | NP_001129079.1 | - | 816 | 106 |
XM_008773382.1 | XP_008771604.1 | - | 617 | 106 | |
XM_008773383.1 | XP_008771605.1 | 4 - 5 | 563 | 106 | |
XM_008773384.1 | XP_008771606.1 | - | 502 | 106 | |
GenBank mRNA | DQ058651.1 | - | - | 816 | 106 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006257477.2 | XP_006257539.1 | 4 - 5 | 563 | 106 |
XM_006257478.2 | XP_006257540.1 | 3 - 4 | 502 | 106 | |
XM_008773510.1 | XP_008771732.1 | 4 - 5 | 617 | 106 |