Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2s1
Entrez Gene ID
308445
Gene Name
cytochrome P450, family 2, subfamily s, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.1: 82595211 - 82610350 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107495.1 NP_001100965.1 1 - 2 230 159

View Full Product Details