Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgma
Entrez Gene ID
308739
Gene Name
repulsive guidance molecule family member A
Gene Aliases
-
Chromosome Location
Chr.1: 134699053 - 134742512 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107524.1 NP_001100994.1 2 - 3 485 121

View Full Product Details