Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btbd1
Entrez Gene ID
293060
Gene Name
BTB (POZ) domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 143609729 - 143647124 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011932.1 NP_001011932.1 7 - 8 1351 70
GenBank mRNA BC085850.1 - 7 - 8 1351 70
FQ221559.1 - 3 - 4 359 70

View Full Product Details