Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Syt8
Entrez Gene ID
60566
Gene Name
synaptotagmin VIII
Gene Aliases
-
Chromosome Location
Chr.1: 215605408 - 215607602 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053325.1 NP_445777.1 1 - 2 124 74
GenBank mRNA AF375462.1 - 1 - 2 124 74
AF375467.1 - 1 - 2 124 74

View Full Product Details