Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dak
Entrez Gene ID
361730
Gene Name
dihydroxyacetone kinase 2 homolog (S. cerevisiae)
Gene Aliases
RGD1311026
Chromosome Location
Chr.1: 226642901 - 226657698 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039031.1 NP_001034120.1 12 - 13 1382 71
XM_006231056.2 XP_006231118.1 12 - 13 1268 71
XM_006231057.2 XP_006231119.2 12 - 13 1551 71
GenBank mRNA BC098925.1 - 12 - 13 1382 71

View Full Product Details