Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcerg1l
Entrez Gene ID
361669
Gene Name
transcription elongation regulator 1-like
Gene Aliases
RGD1562437
Chromosome Location
Chr.1: 210550668 - 210739600 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130077.1 NP_001123549.1 10 - 11 1410 153

View Full Product Details