Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ache
Entrez Gene ID
83817
Gene Name
acetylcholinesterase
Gene Aliases
-
Chromosome Location
Chr.12: 22472358 - 22477052 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC094521.1 - - 1648 136

View Full Product Details