Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb37
Entrez Gene ID
304918
Gene Name
zinc finger and BTB domain containing 37
Gene Aliases
RGD1308731
Chromosome Location
Chr.13: 78836491 - 78852211 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107191.1 NP_001100661.1 2 - 3 230 98

View Full Product Details