The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107191.1 | NP_001100661.1 | 4 - 5 | 1289 | 68 |
XM_006250110.2 | XP_006250172.1 | 3 - 4 | 1144 | 68 | |
XM_006250111.2 | XP_006250173.1 | 3 - 4 | 1147 | 68 | |
XM_008769648.1 | XP_008767870.1 | 4 - 5 | 1337 | 68 |