Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pter
Entrez Gene ID
63852
Gene Name
phosphotriesterase related
Gene Aliases
Rpr-1, Rpr1
Chromosome Location
Chr.17: 80250443 - 80311987 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254291.2 XP_006254353.1 1 - 2 123 105

View Full Product Details