Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm6
Entrez Gene ID
307305
Gene Name
PR domain containing 6
Gene Aliases
RGD1559594
Chromosome Location
Chr.18: 48433747 - 48538834 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772191.1 XP_008770413.1 7 - 8 1754 86

View Full Product Details