Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc21a4
- Entrez Gene ID
-
80899
- Gene Name
- kidney specific organic anion transporter
- Gene Aliases
-
OAT-K1, OAT-K2, Slco1a3, rOAT-K1, rOAT-K11, rOAT-K13, rOAT-K14, rOAT-K2, rOAT-K3, rOAT-K5, rOAT-K6, rOAT-K7, rOAT-K8, rOAT-K9
- Chromosome Location
-
Chr.4: 176255603 - 176299119 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB012662.1 |
- |
2 - 3 |
137 |
100 |
View Full Product Details