Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grcc10
Entrez Gene ID
100362005
Gene Name
gene rich cluster, C10 gene
Gene Aliases
-
Chromosome Location
Chr.4: 157264383 - 157266039 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198725.1 NP_001185654.1 2 - 3 528 114
GenBank mRNA FM062242.1 - 2 - 3 307 114

Gene Symbol
LOC100911713
Entrez Gene ID
100911713
Gene Name
protein C10-like
Gene Aliases
-
Chromosome Location
Chr.9: 20279612 - 20281274 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750659.3 XP_003750707.1 2 - 3 550 114

View Full Product Details