The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105756.1 | NP_001099226.1 | 1 - 1 | 349 | 114 |
GenBank mRNA | AF226629.1 | - | 1 - 1 | 295 | 114 |