Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chodl
Entrez Gene ID
288289
Gene Name
chondrolectin
Gene Aliases
-
Chromosome Location
Chr.11: 17538009 - 17560305 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105894.1 NP_001099364.1 2 - 3 830 99
XM_006248003.2 XP_006248065.1 2 - 3 884 99
XM_006248004.2 XP_006248066.1 2 - 3 812 99

View Full Product Details