Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sssca1
Entrez Gene ID
689397
Gene Name
Sjogren syndrome/scleroderma autoantigen 1
Gene Aliases
-
Chromosome Location
Chr.1: 221087803 - 221089586 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109537.2 NP_001103007.1 2 - 3 272 83

View Full Product Details