The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108094.2 | NP_001101564.1 | 9 - 10 | 1149 | 106 |
GenBank mRNA | BC128768.1 | - | 9 - 10 | 1149 | 106 |