Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a32
Entrez Gene ID
315023
Gene Name
solute carrier family 25 (mitochondrial folate carrier) , member 32
Gene Aliases
RGD1565789
Chromosome Location
Chr.7: 77983885 - 78005054 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173334.1 NP_001166805.1 2 - 3 485 136
XM_006241603.2 XP_006241665.1 2 - 3 564 136
GenBank mRNA BC166530.1 - 2 - 3 485 136

View Full Product Details