Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a32
Entrez Gene ID
315023
Gene Name
solute carrier family 25 (mitochondrial folate carrier) , member 32
Gene Aliases
RGD1565789
Chromosome Location
Chr.7: 77983885 - 78005054 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173334.1 NP_001166805.1 6 - 7 989 113
GenBank mRNA BC166530.1 - 6 - 7 989 113

View Full Product Details