Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
A4galt
Entrez Gene ID
63888
Gene Name
alpha 1,4-galactosyltransferase
Gene Aliases
Gb3
Chromosome Location
Chr.7: 124085832 - 124113396 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242118.2 XP_006242180.1 1 - 2 504 57

View Full Product Details