Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptp4a3
Entrez Gene ID
362930
Gene Name
protein tyrosine phosphatase type IVA, member 3
Gene Aliases
-
Chromosome Location
Chr.7: 114971214 - 115002902 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114405.2 NP_001107877.1 4 - 5 900 79
XM_006241729.2 XP_006241791.1 6 - 7 1397 79
XM_006241731.2 XP_006241793.1 5 - 6 2507 79
XM_006241732.2 XP_006241794.1 5 - 6 1319 79
XM_006241733.2 XP_006241795.1 5 - 6 1278 79
XM_008765573.1 XP_008763795.1 6 - 7 2585 79
XM_008765574.1 XP_008763796.1 5 - 6 1964 79
GenBank mRNA BC159432.1 - 4 - 5 900 79

View Full Product Details