Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pgm2l1
Entrez Gene ID
685076
Gene Name
phosphoglucomutase 2-like 1
Gene Aliases
-
Chromosome Location
Chr.1: 165237847 - 165286980 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109454.1 NP_001102924.1 1 - 2 417 96
XM_006229793.2 XP_006229855.1 1 - 2 417 96

View Full Product Details