Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfp46
Entrez Gene ID
298558
Gene Name
zinc finger protein 46
Gene Aliases
-
Chromosome Location
Chr.5: 154644096 - 154655132 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008764219.1 XP_008762441.1 1 - 2 414 68

View Full Product Details