Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lin52
Entrez Gene ID
362763
Gene Name
lin-52 DREAM MuvB core complex component
Gene Aliases
RGD1306798
Chromosome Location
Chr.6: 108167247 - 108264445 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240356.2 XP_006240418.1 2 - 3 573 72
XM_006240358.2 XP_006240420.1 2 - 3 399 72
XM_008764889.1 XP_008763111.1 2 - 3 396 72
XM_008764890.1 XP_008763112.1 2 - 3 389 72
XM_343089.7 XP_343090.4 2 - 3 385 72

View Full Product Details