Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psma5
Entrez Gene ID
29672
Gene Name
proteasome (prosome, macropain) subunit, alpha type 5
Gene Aliases
-
Chromosome Location
Chr.2: 211050385 - 211073703 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017282.1 NP_058978.1 3 - 4 312 110
GenBank mRNA BC060575.1 - 3 - 4 333 110
D10756.1 - 3 - 4 312 110

View Full Product Details