Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfkbil1
Entrez Gene ID
361794
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1
Gene Aliases
-
Chromosome Location
Chr.20: 4823612 - 4839202 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212509.2 NP_997674.1 2 - 3 445 261
XM_006256046.2 XP_006256108.1 2 - 3 380 261
GenBank mRNA BC099772.1 - 2 - 3 389 261

View Full Product Details