Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sult1c3
- Entrez Gene ID
-
65185
- Gene Name
- sulfotransferase family, cytosolic, 1C, member 3
- Gene Aliases
-
HAST-I, St1c1, Stp2, Sult1a2, Sult1c1
- Chromosome Location
-
Chr.9: 4931038 - 4978847 on Build Rnor_6.0
- Gene Symbol
- LOC100910235
- Entrez Gene ID
-
100910235
- Gene Name
- sulfotransferase 1C1-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 4323997 - 4328184 on Build Rnor_6.0
View Full Product Details