Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cmah
Entrez Gene ID
361245
Gene Name
cytidine monophospho-N-acetylneuraminic acid hydroxylase
Gene Aliases
Cmahp
Chromosome Location
Chr.17: 42583939 - 42640221 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024273.1 NP_001019444.1 13 - 14 1733 104
GenBank mRNA BC095843.1 - 13 - 14 1733 104

View Full Product Details