Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hoxc4
Entrez Gene ID
24459
Gene Name
homeo box C4
Gene Aliases
Hox3r3
Chromosome Location
Chr.7: 144646608 - 144650129 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109884.1 NP_001103354.1 1 - 2 865 73
XM_008765667.1 XP_008763889.1 2 - 3 1695 73

View Full Product Details