The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106770.1 | NP_001100240.1 | 2 - 3 | 316 | 87 |
GenBank mRNA | FQ211295.1 | - | 2 - 3 | 309 | 87 |
FQ211839.1 | - | 2 - 3 | 299 | 87 | |
FQ217217.1 | - | 2 - 3 | 299 | 87 | |
FQ221492.1 | - | 2 - 3 | 303 | 87 | |
FQ224331.1 | - | 2 - 3 | 303 | 87 | |
FQ224393.1 | - | 2 - 3 | 87 | ||
FQ224740.1 | - | 2 - 3 | 301 | 87 |