The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013188.1 | NP_037320.1 | 17 - 18 | 2292 | 62 |
GenBank mRNA | BC166475.1 | - | - | 2248 | 62 |
CO400919.1 | - | 3 - 4 | 417 | 62 |